accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClICl

CAS RN 2401-21-0

IUPAC Name: 1,2-dichloro-3-iodobenzene
Synonyms: 1,2-dichloro-3-iodobenzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: VGJKBWPZBVBXGI-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(I)=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results