accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClH2N

CAS RN 2401-24-3

IUPAC Name: 2-chloro-5-methoxyaniline
Synonyms: 2-chloro-5-methoxyaniline
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: GBOUQGUQUUPGLO-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(N)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)