CAS RN 2404-44-6
IUPAC Name:
2-octyloxirane
Synonyms:
2-octyloxirane
Molecular Weight (g/mol):
156.27
Molecular Formula:
C10H20O
InChi Key:
AAMHBRRZYSORSH-UHFFFAOYNA-N
SMILES:
CCCCCCCCC1CO1
1
–
1
of
1
results
1
1,2-Epoxydecane, 97%
CAS: 2404-44-6 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 MDL Number: MFCD00005158 InChI Key: AAMHBRRZYSORSH-UHFFFAOYNA-N Synonym: 1,2-epoxydecane,oxirane, octyl,octyloxirane,oxirane, 2-octyl,1,2-decylene oxide,ccris 2615,epoxides, c>8-alkyl,decene oxide,epoxy decane,oxirane,octyl PubChem CID: 16993 IUPAC Name: 2-octyloxirane SMILES: CCCCCCCCC1CO1