accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOHOON

CAS RN 24057-28-1

IUPAC Name: 4-methylbenzene-1-sulfonic acid; pyridine
Synonyms: pyridinium p-toluenesulfonate
Molecular Weight (g/mol): 251.3
Molecular Formula: C12H13NO3S
InChi Key: ZDYVRSLAEXCVBX-UHFFFAOYSA-N
SMILES: C1=CC=NC=C1.CC1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)