accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24067-17-2

IUPAC Name: (4-nitrophenyl)boronic acid
Synonyms: 4-nitrophenylboronic acid
Molecular Weight (g/mol): 166.93
Molecular Formula: C6H6BNO4
InChi Key: NSFJAFZHYOAMHL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results