accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2409-61-2

IUPAC Name: bis(4-methylphenyl)(oxo)-λ⁵-phosphanylium
Synonyms: bis(4-methylphenyl)(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 229.24
Molecular Formula: C14H14OP
InChi Key: ZHIPXAFNKGZMSC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[P+](=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results