CAS RN 241496-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-3-(4-bromobenzoyl)benzoic Acid, TRC
CAS: 241496-82-2 Molecular Formula: C14 H10 Br N O3 Molecular Weight (g/mol): 320.14 IUPAC Name: 2-amino-3-(4-bromobenzoyl)benzoic acid SMILES: Nc1c(cccc1C(=O)c2ccc(Br)cc2)C(=O)O