CAS RN 2415-80-7
IUPAC Name:
(2,2-dichlorocyclopropyl)benzene
Synonyms:
(2,2-dichlorocyclopropyl)benzene
Molecular Weight (g/mol):
187.06
Molecular Formula:
C9H8Cl2
InChi Key:
WLWFQGXZIDYWQF-UHFFFAOYNA-N
SMILES:
ClC1(Cl)CC1C1=CC=CC=C1
1
–
1
of
1
results
1
1,1-Dichloro-2-phenylcyclopropane, 97%, Thermo Scientific™
CAS: 2415-80-7 Molecular Formula: C9H8Cl2 Molecular Weight (g/mol): 187.063 MDL Number: MFCD00001272 InChI Key: WLWFQGXZIDYWQF-UHFFFAOYSA-N Synonym: 2,2-dichlorocyclopropyl benzene,1,1-dichloro-2-phenylcyclopropane,2,2-dichlorocyclopropyl-benzene,benzene, 2,2-dichlorocyclopropyl,cyclopropane, 1,1-dichloro, 2-phenyl,pubchem18679,acmc-1clka,ksc491m2n,wlwfqgxzidywqf-uhfffaoysa,2,2-dichloro-cyclopropyl-benzene PubChem CID: 95349 IUPAC Name: (2,2-dichlorocyclopropyl)benzene SMILES: C1C(C1(Cl)Cl)C2=CC=CC=C2