accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2416-94-6

CH3CH3HOH3C

CAS RN 2416-94-6

IUPAC Name: 2,3,6-trimethylphenol
Synonyms: 2,3,6-trimethylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: QQOMQLYQAXGHSU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results