accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2416-95-7

IUPAC Name: 3,3',5,5'-tetrakis(propan-2-yl)-[1,1'-biphenyl]-4,4'-diol
Synonyms: 3,3',5,5'-tetraisopropyl-[1,1'-biphenyl]-4,4'-diol
Molecular Weight (g/mol): 354.53
Molecular Formula: C24H34O2
InChi Key: QAISRHCMPQROAX-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=CC(C(C)C)=C1O)C1=CC(C(C)C)=C(O)C(=C1)C(C)C
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results