accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOOClONNClCl

CAS RN 24169-02-6

IUPAC Name: 1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole; nitric acid
Synonyms: econazole; nitric acid
Molecular Weight (g/mol): 444.69
Molecular Formula: C18H16Cl3N3O4
InChi Key: DDXORDQKGIZAME-UHFFFAOYNA-N
SMILES: O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results