accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2420-94-2

IUPAC Name: hydrogen 2-aminoethyl 2-methylprop-2-enoate chloride
Synonyms: hydrogen 2-aminoethyl 2-methylprop-2-enoate chloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: XSHISXQEKIKSGC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(=C)C(=O)OCCN
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results