CAS RN 24255-97-8
CAS RN 24255-97-8
IUPAC Name:
2-bromo-6-(piperidin-1-yl)pyridine
Synonyms:
2-bromo-6-(piperidin-1-yl)pyridine
Molecular Weight (g/mol):
241.13
Molecular Formula:
C10H13BrN2
InChi Key:
YHQZMCNKHWPFBS-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=N1)N1CCCCC1
1
–
1
of
1
results
1
2-Bromo-6-piperidinopyridine, ≥97%, Thermo Scientific™
CAS: 24255-97-8 Molecular Formula: C10H13BrN2 Molecular Weight (g/mol): 241.132 MDL Number: MFCD07772823 InChI Key: YHQZMCNKHWPFBS-UHFFFAOYSA-N Synonym: 2-bromo-6-piperidin-1-yl pyridine,2-bromo-6-piperidinopyridine,acmc-1cmoy,6-bromo-2-piperidylpyridine,2-bromo-6-1-piperidinyl pyridine,2-bromo-6-piperadinopyridine,2-bromanyl-6-piperidin-1-yl-pyridine,pyridine,2-bromo-6-1-piperidinyl,6'-bromo-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl PubChem CID: 7164597 IUPAC Name: 2-bromo-6-piperidin-1-ylpyridine SMILES: C1CCN(CC1)C2=NC(=CC=C2)Br