accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24259-59-4

(R)(S)(S)OOHHOOHHO

CAS RN 24259-59-4

IUPAC Name: (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
Synonyms: L-ribose aldehydo-L-ribose aldehydo-L-ribo-pentose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: PYMYPHUHKUWMLA-MROZADKFSA-N
SMILES: OC[C@H](O)[C@H](O)[C@H](O)C=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)