accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24304-84-5

IUPAC Name: 2-[(2-aminoethyl)sulfanyl]ethan-1-ol
Synonyms: 2-[(2-aminoethyl)sulfanyl]ethanol
Molecular Weight (g/mol): 121.2
Molecular Formula: C4H11NOS
InChi Key: ZRZOERYTFAWUQQ-UHFFFAOYSA-N
SMILES: NCCSCCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results