accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 2433-97-8

IUPAC Name: methyl tricosanoate
Synonyms: methyl tricosanoate
Molecular Weight (g/mol): 368.65
Molecular Formula: C24H48O2
InChi Key: VORKGRIRMPBCCZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)