accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOOH

CAS RN 2437-17-4

IUPAC Name: 6-hydroxynaphthalene-1-carboxylic acid
Synonyms: 6-hydroxynaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: JCJUKCIXTRWAQY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC(O)=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)