accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24372-48-3

OSN

CAS RN 24372-48-3

IUPAC Name: 5-(piperidin-1-yl)thiophene-2-carbaldehyde
Synonyms: 5-(piperidin-1-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 195.28
Molecular Formula: C10H13NOS
InChi Key: SFDXRMCPPLZJBG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results