CAS RN 24386-93-4
IUPAC Name:
2-{4-amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms:
2-{4-amino-5-iodopyrrolo[2,3-d]pyrimidin-7-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol):
392.15
Molecular Formula:
C11H13IN4O4
InChi Key:
WHSIXKUPQCKWBY-UHFFFAOYNA-N
SMILES:
NC1=C2C(I)=CN(C3OC(CO)C(O)C3O)C2=NC=N1
1
–
2
of
2
results
1
5-Iodotubercidin, TRC
CAS: 24386-93-4 Molecular Formula: C11 H13 I N4 O4 Molecular Weight (g/mol): 392.15 IUPAC Name: (2R,3R,4S,5R)-2-(4-amino-5-iodopyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: Nc1ncnc2c1c(I)cn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
2
Promotions Available
Tocris Bioscience™ 5-Iodotubercidin