accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 243863-36-7

NH2OFF

CAS RN 243863-36-7

IUPAC Name: 1-[2-(difluoromethoxy)phenyl]methanamine
Synonyms: 1-[2-(difluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: GGXXUJUQXAOYPK-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1OC(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results