accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2439-35-2

IUPAC Name: 2-(dimethylamino)ethyl prop-2-enoate
Synonyms: 2-(dimethylamino)ethyl prop-2-enoate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: DPBJAVGHACCNRL-UHFFFAOYSA-N
SMILES: CN(C)CCOC(=O)C=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results