CAS RN 244101-02-8
IUPAC Name:
N-(5-bromo-2-methoxybenzenesulfonyl)-3-{2-[(naphthalen-2-yl)methyl]phenyl}prop-2-enamide
Synonyms:
N-(5-bromo-2-methoxybenzenesulfonyl)-3-[2-(naphthalen-2-ylmethyl)phenyl]prop-2-enamide
Molecular Weight (g/mol):
536.44
Molecular Formula:
C27H22BrNO4S
InChi Key:
ODTKFNUPVBULRJ-UHFFFAOYSA-N
SMILES:
COC1=C(C=C(Br)C=C1)S(=O)(=O)NC(=O)C=CC1=CC=CC=C1CC1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1