accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHBr

CAS RN 244205-40-1

IUPAC Name: (2-bromophenyl)boronic acid
Synonyms: 2-bromophenylboronic acid
Molecular Weight (g/mol): 200.83
Molecular Formula: C6H6BBrO2
InChi Key: PLVCYMZAEQRYHJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results