accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24424-99-5

IUPAC Name: di-tert-butyl dicarbonate
Synonyms: di-tert-butyl dicarbonate tert-butyl dicarbonate boc2O boc(2)o cpd di-tert-butyldicarbonate boc-anhydride boc(2)O cpd boc2O cpd bis(tert-butoxycarbonyl)oxide tert-butoxycarbonyl anhydride
Molecular Weight (g/mol): 218.25
Molecular Formula: C10H18O5
InChi Key: DYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (6)
  • (12)
Quantity 
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Boiling Point 
  • (6)
  • (7)
Percent Purity 
  • (6)
  • (2)
  • (5)
Physical Form 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (12)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (6)
  • (7)
  • (6)
  • (2)
  • (5)
  • (2)
  • (7)