accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24440-15-1

NHOH2NOH(E/Z)

CAS RN 24440-15-1

IUPAC Name: N',2-dihydroxy-2-phenylethanimidamide
Synonyms: N',2-dihydroxy-2-phenylethanimidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: GXYVISXLFBQBKH-UHFFFAOYNA-N
SMILES: NC(=NO)C(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results