accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24443-15-0

CH3CH3BrBr

CAS RN 24443-15-0

IUPAC Name: 1,3-dibromo-3-methylbutane
Synonyms: 1,3-dibromo-3-methylbutane
Molecular Weight (g/mol): 229.94
Molecular Formula: C5H10Br2
InChi Key: XMJFMQSXQQUJTQ-UHFFFAOYSA-N
SMILES: CC(C)(Br)CCBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results