accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24451-15-8

IUPAC Name: ethyl [(4-bromophenyl)carbamoyl]formate
Synonyms: ethyl [(4-bromophenyl)carbamoyl]formate
Molecular Weight (g/mol): 272.1
Molecular Formula: C10H10BrNO3
InChi Key: RWWFLEVLFXJHIB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)NC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results