accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH

CAS RN 24463-15-8

IUPAC Name: (pyren-1-yl)methanol
Synonyms: pyren-1-ylmethanol
Molecular Weight (g/mol): 232.28
Molecular Formula: C17H12O
InChi Key: NGDMLQSGYUCLDC-UHFFFAOYSA-N
SMILES: OCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results