accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24463-19-2

IUPAC Name: 9-(chloromethyl)anthracene
Synonyms: 9-chloromethylanthracene
Molecular Weight (g/mol): 226.7
Molecular Formula: C15H11Cl
InChi Key: PCVRSXXPGXRVEZ-UHFFFAOYSA-N
SMILES: ClCC1=C2C=CC=CC2=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)