accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBOHOH

CAS RN 24464-63-9

IUPAC Name: decylboronic acid
Synonyms: decylboronic acid
Molecular Weight (g/mol): 186.1
Molecular Formula: C10H23BO2
InChi Key: TUTSEMMBKGBUGD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCB(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results