CAS RN 244767-67-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dapivirine, TRC
CAS: 244767-67-7 Molecular Formula: C20 H19 N5 Molecular Weight (g/mol): 329.4 Synonym: Benzonitrile, 4-[[4-[(2,4,6-trimethylphenyl)amino]-2-pyrimidinyl]amino]-,4-[[4-[(2,4,6-Trimethylphenyl)amino]-2-pyrimidinyl]amino]benzonitrile,AIDS 105293,Dapivirine,R 147681,TMC 120,TMC 120R147681 IUPAC Name: 4-[[4-(2,4,6-trimethylanilino)pyrimidin-2-yl]amino]benzonitrile SMILES: Cc1cc(C)c(Nc2ccnc(Nc3ccc(cc3)C#N)n2)c(C)c1