accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2454-39-9

IUPAC Name: 2-aminobenzene-1-carbothioamide
Synonyms: O-aminothiobenzamide
Molecular Weight (g/mol): 152.22
Molecular Formula: C7H8N2S
InChi Key: HPZKAJRFABCGFF-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results