accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2455-14-3

CH3H3CCH3OH3CH3CCH3OCH3H3CCH3CH3CH3H3C

CAS RN 2455-14-3

IUPAC Name: 3,3',5,5'-tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Synonyms: 3,3',5,5'-tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Molecular Weight (g/mol): 408.63
Molecular Formula: C28H40O2
InChi Key: GQIGHOCYKUBBOE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C=C(C1=O)C(C)(C)C)=C1C=C(C(=O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)