accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNOO

CAS RN 24566-79-8

IUPAC Name: 2-(6-bromohexyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(6-bromohexyl)isoindole-1,3-dione
Molecular Weight (g/mol): 310.19
Molecular Formula: C14H16BrNO2
InChi Key: OAZFTIPKNPTDIO-UHFFFAOYSA-N
SMILES: BrCCCCCCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results