accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONCH3H3CCH3OHI

CAS RN 24570-49-8

IUPAC Name: [(4-hydroxy-5-methyloxolan-2-yl)methyl]trimethylazanium iodide
Synonyms: [(4-hydroxy-5-methyloxolan-2-yl)methyl]trimethylazanium iodide
Molecular Weight (g/mol): 301.17
Molecular Formula: C9H20INO2
InChi Key: PMFYONXEPDMBPE-UHFFFAOYNA-M
SMILES: [I-].CC1OC(C[N+](C)(C)C)CC1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results