accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2NOHO

CAS RN 2458-12-0

IUPAC Name: 3-amino-4-methylbenzoic acid
Synonyms: 3-amino-4-methylbenzoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: XKFIFYROMAAUDL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results