CAS RN 24584-09-6
CAS RN 24584-09-6
IUPAC Name:
4-[(2S)-2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione
Synonyms:
dexrazoxane dexrazoxano [inn-spanish] (+)-dexrazoxane dexrazoxano dextrorazoxane razoxana [inn-spanish] dexrazoxanum razoxanum desrazoxane
Molecular Weight (g/mol):
268.27
Molecular Formula:
C11H16N4O4
InChi Key:
BMKDZUISNHGIBY-ZETCQYMHSA-N
SMILES:
C[C@@H](CN1CC(=O)NC(=O)C1)N1CC(=O)NC(=O)C1
1
–
1
of
1
results
1
Dexrazoxane, TRC
CAS: 24584-09-6 Molecular Formula: C11 H16 N4 O4 Molecular Weight (g/mol): 268.27 Synonym: 2,6-Piperazinedione, 4,4'-[(1S)-1-methyl-1,2-ethanediyl]bis-,2,6-Piperazinedione, 4,4'-(1-methyl-1,2-ethanediyl)bis-, (S)-,2,6-Piperazinedione, 4,4'-propylenedi-, (+)- (8CI),4,4'-[(1S)-1-Methyl-1,2-ethanediyl]bis[2,6-piperazinedione],(+)-1,2-Bis(3,5-dioxopiperazin-1-yl)propane,(S)-(+)-1,2-Bis(3,5-dioxopiperazin-1-yl)propane,ADR 529,Dexrazoxane,ICRF 187,NSC 169780,Savene,Totect,Zinecard IUPAC Name: 4-[(2S)-2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione SMILES: C[C@@H](CN1CC(=O)NC(=O)C1)N2CC(=O)NC(=O)C2