accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24589-77-3

IUPAC Name: hydrogen 4-hydrazinylbenzoic acid chloride
Synonyms: hydrogen 4-hydrazinylbenzoic acid chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: XHLQMKQBCHYRLC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results