accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOFFFSiCH3CH3H3C

CAS RN 24589-78-4

IUPAC Name: 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Synonyms: 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Molecular Weight (g/mol): 199.25
Molecular Formula: C6H12F3NOSi
InChi Key: MSPCIZMDDUQPGJ-UHFFFAOYSA-N
SMILES: CN(C(=O)C(F)(F)F)[Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results