accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24596-19-8

IUPAC Name: 4-bromo-2,6-dimethylaniline
Synonyms: 4-bromo-2,6-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: QGLAYJCJLHNIGJ-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(C)=C1N
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results