accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KOOONHOHN

CAS RN 24598-73-0

IUPAC Name: potassium 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Synonyms: potassium orotate
Molecular Weight (g/mol): 194.19
Molecular Formula: C5H3KN2O4
InChi Key: DHBUISJCVRMTAZ-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C1=CC(=O)NC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results