accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2460-59-5

IUPAC Name: 2-formyl-4,6-dinitrobenzen-1-olate
Synonyms: 2-formyl-4,6-dinitrobenzenolate
Molecular Weight (g/mol): 211.11
Molecular Formula: C7H3N2O6
InChi Key: FLJXIBHYDIMYRS-UHFFFAOYSA-M
SMILES: [O-]C1=C(C=O)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results