accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2460-87-9

IUPAC Name: 4-(tert-butoxy)phenol
Synonyms: 4-(tert-butoxy)phenol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: CIICLJLSRUHUBY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results