accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24603-63-2

IUPAC Name: 5-chloro-2,3-dihydro-1,2-benzoxazol-3-one
Synonyms: 5-chloro-2H-1,2-benzoxazol-3-one
Molecular Weight (g/mol): 169.56
Molecular Formula: C7H4ClNO2
InChi Key: WIZWHBRFTCYPDN-UHFFFAOYSA-N
SMILES: ClC1=CC=C2ONC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results