accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH2NOO

CAS RN 2462-31-9

IUPAC Name: benzyl 2-aminoacetate hydrochloridyl
Synonyms: benzyl 2-aminoacetate hydrochloridyl
Molecular Weight (g/mol): 200.64
Molecular Formula: C9H11ClNO2
InChi Key: BIGFBRPKXPVZGE-UHFFFAOYSA-N
SMILES: [Cl].NCC(=O)OCC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)