accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2462-84-2

CH3OOCH3(E)

CAS RN 2462-84-2

IUPAC Name: methyl (9E)-octadec-9-enoate
Synonyms: methyl oleate emery, oleic acid ester oleic acid ester emery Edenor MeTiO5 phytorob 926-67 (E)-methyl oleate (e)-methyl oleate edenor me 90/95V exceparl M-ol emery oleic acid ester 2301
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results