accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OHO

CAS RN 24623-65-2

IUPAC Name: 3-tert-butyl-2-hydroxybenzaldehyde
Synonyms: 3-tert-butyl-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: ROILLNJICXGZQQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC(C=O)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results