accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24634-61-5

IUPAC Name: potassium (2E,4E)-hexa-2,4-dienoate
Synonyms: potassium sorbate
Molecular Weight (g/mol): 150.22
Molecular Formula: C6H7KO2
InChi Key: CHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES: [K+].C\C=C\C=C\C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results