accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3O

CAS RN 24650-42-8

IUPAC Name: 2,2-dimethoxy-1,2-diphenylethan-1-one
Synonyms: phenyldimethoxyacetophenone
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: KWVGIHKZDCUPEU-UHFFFAOYSA-N
SMILES: COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)