accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24683-00-9

CH3ONNH3CCH3

CAS RN 24683-00-9

IUPAC Name: 2-methoxy-3-(2-methylpropyl)pyrazine
Synonyms: isobutyl-methoxypyrazine
Molecular Weight (g/mol): 166.22
Molecular Formula: C9H14N2O
InChi Key: UXFSPRAGHGMRSQ-UHFFFAOYSA-N
SMILES: COC1=NC=CN=C1CC(C)C
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)